CAS RN 6087-13-4
IUPAC Name:
2-fluoropropanoic acid
Synonyms:
2-fluoropropanoic acid
Molecular Weight (g/mol):
92.07
Molecular Formula:
C3H5FO2
InChi Key:
ZVZPFTCEXIGSHM-UHFFFAOYNA-N
SMILES:
CC(F)C(O)=O
1
–
1
of
1
results
1
Promotions Available
2-Fluoropropionic acid, 96%, Thermo Scientific Chemicals
2-Fluoropropionic acid, 96%, Thermo Scientific Chemicals
CAS: 6087-13-4 Molecular Formula: C3H5FO2 Molecular Weight (g/mol): 92.069 MDL Number: MFCD06247714 InChI Key: ZVZPFTCEXIGSHM-UHFFFAOYSA-N Synonym: 2-fluoropropionic acid,2-fluoro-propionic acid,propanoic acid, 2-fluoro,pubchem13698,propanoic acid,2-fluoro,acmc-1b20x,ksc492i2p PubChem CID: 256876 IUPAC Name: 2-fluoropropanoic acid SMILES: CC(C(=O)O)F