missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoropropionic acid, 96%, Thermo Scientific Chemicals

Catalog No. AAH2584103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2584103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2584103 Supplier Thermo Scientific Chemicals Supplier No. H2584103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6087-13-4 | Molecular Formula: C3H5FO2 | Molecular Weight: 92.069 g/mol

2-Fluoropropionic acid is used in the medical field for diagnostic purposes. It is labeled with 18F and is used as an imaging agent for detecting prostate cancer in humans. It is also used to prepare 2-fluoro fatty acids, compounds that exhibit antifungal properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoropropionic acid is used in the medical field for diagnostic purposes. It is labeled with 18F and is used as an imaging agent for detecting prostate cancer in humans. It is also used to prepare 2-fluoro fatty acids, compounds that exhibit antifungal properties.

Solubility
Partly miscible in water.

Notes
Store in a cool, dry place in tightly closed container. Store away from oxidizing agents, strong bases.

Chemical Identifiers

CAS 6087-13-4
Molecular Formula C3H5FO2
Molecular Weight (g/mol) 92.069
MDL Number MFCD06247714
InChI Key ZVZPFTCEXIGSHM-UHFFFAOYSA-N
Synonym 2-fluoropropionic acid, 2-fluoro-propionic acid, propanoic acid, 2-fluoro, pubchem13698, propanoic acid,2-fluoro, acmc-1b20x, ksc492i2p
PubChem CID 256876
IUPAC Name 2-fluoropropanoic acid
SMILES CC(C(=O)O)F

Specifications

Density 1.214
Boiling Point 66°C to 67°C (30 mmHg)
Flash Point 82°C (180°F)
Odor Acrid
Refractive Index 1.383
Quantity 1 g
UN Number UN3265
Solubility Information Partly miscible in water.
Formula Weight 92.07
Percent Purity 96%
Chemical Name or Material 2-Fluoropropionic acid
Show More Show Less
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only