accessibility menu, dialog, popup

UserName

NH2NOONOO

CAS RN 606-22-4

IUPAC Name: 2,6-dinitroaniline
Synonyms: 2,6-dinitroaniline
Molecular Weight (g/mol): 183.12
Molecular Formula: C6H5N3O4
InChi Key: QFUSCYRJMXLNRB-UHFFFAOYSA-N
SMILES: NC1=C(C=CC=C1[N+]([O-])=O)[N+]([O-])=O
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)