accessibility menu, dialog, popup

UserName

CAS RN 6055-19-2

IUPAC Name: 2-[bis(2-chloroethyl)amino]-1,3,2λ⁵-oxazaphosphinan-2-one hydrate
Synonyms: cyclophosphamide monohydrate cyclophosphamide hydrate
Molecular Weight (g/mol): 279.1
Molecular Formula: C7H17Cl2N2O3P
InChi Key: PWOQRKCAHTVFLB-UHFFFAOYNA-N
SMILES: O.ClCCN(CCCl)P1(=O)NCCCO1
Percent Purity 
  • (1)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Grade 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)