accessibility menu, dialog, popup

UserName

PbOOO

CAS RN 598-63-0

IUPAC Name: λ²-lead(2+) carbonate
Synonyms: lead carbonate
Molecular Weight (g/mol): 267.2
Molecular Formula: CO3Pb
InChi Key: MFEVGQHCNVXMER-UHFFFAOYSA-L
SMILES: [Pb++].[O-]C([O-])=O
Quantity 
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
Grade 
  • (4)
  • (1)
  • (6)
  • (4)
Physical Form 
  • (6)
  • (4)
  • (4)
  • (1)
Molecular Weight (g/mol) 
  • (12)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (10)
  • (10)
  • (1)
  • (10)
  • (5)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (4)
  • (1)
  • (6)
  • (4)
  • (6)
  • (4)
  • (4)
  • (1)
  • (12)
  • (2)
  • (4)