missing translation for 'onlineSavingsMsg'
Learn More

Lead Carbonate, Powder, Reagent, ACS, Spectrum™ Chemical

Catalog No. 18603766
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
6 x 500 g
12 kg
50 kg
Packaging:
Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603766 50 kg Poly Bottle
18603761 125 g Poly Bottle
18603762 500 g Poly Bottle
18603764 2.5 kg Poly Bottle
18603763 6 x 500 g Poly Bottle
18603765 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18603766 Supplier Spectrum Chemical Mfg Cor Supplier No. L104550KGBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

L1045, 598-63-0, PbCO3

Spectrum™ Chemical Lead Carbonate, Powder, Reagent, ACS is a chemical compound that is created industrially from lead acetate and carbon dioxide. It is an important pigment used in paints, putty, and ceramics. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 598-63-0
Molecular Formula CO3Pb
Molecular Weight (g/mol) 267.20
InChI Key MFEVGQHCNVXMER-UHFFFAOYSA-L
IUPAC Name λ2-lead(2+) carbonate
SMILES [Pb++].[O-]C([O-])=O

Specifications

CAS Max % 100%
Physical Form Crystalline Powder or Needles
Quantity 50 kg
Grade ACS/Reagent
Packaging Poly Bottle
Chemical Name or Material Lead Carbonate, Powder