missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead Carbonate, Powder, CP, Approx. 95%, Spectrum™ Chemical
Specifications
Specifications
| Physical Form | Crystalline Powder, Needles or Crystals |
| CAS Max % | 1 |
| Quantity | 50 kg |
| Packaging | Poly Pail |
| Percent Purity | ∼95% |
| Grade | Not Available |
| Chemical Name or Material | Lead Carbonate, Powder |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.