accessibility menu, dialog, popup

UserName

CAS RN 5959-56-8

IUPAC Name: 4-hydroxynaphthalen-1-aminium chloride
Synonyms: 4-hydroxynaphthalen-1-aminium chloride
Molecular Weight (g/mol): 195.65
Molecular Formula: C10H10ClNO
InChi Key: FDBQTRARWCKEJY-UHFFFAOYSA-N
SMILES: [Cl-].[NH3+]C1=C2C=CC=CC2=C(O)C=C1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (1)
  • (2)
  • (3)
Grade 
  • (5)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (4)
  • (2)
  • (1)
  • (5)
  • (1)
  • (2)
  • (3)
  • (5)
  • (1)
  • (2)