accessibility menu, dialog, popup

UserName

CAS RN 59414-23-2

CH3OCH2OSiH3CH3CCH3(Z)

CAS RN 59414-23-2

IUPAC Name: {[(3Z)-4-methoxybuta-1,3-dien-2-yl]oxy}trimethylsilane
Synonyms: {[(3Z)-4-methoxybuta-1,3-dien-2-yl]oxy}trimethylsilane
Molecular Weight (g/mol): 172.3
Molecular Formula: C8H16O2Si
InChi Key: SHALBPKEGDBVKK-SREVYHEPSA-N
SMILES: CO\C=C/C(=C)O[Si](C)(C)C
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)