missing translation for 'onlineSavingsMsg'
Learn More

1-Methoxy-3-(trimethylsilyloxy)-1,3-butadiene, 90%

Catalog No. AC199550050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC199550050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC199550050 Supplier Thermo Scientific Chemicals Supplier No. 199550050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 59414-23-2 | C8H16O2Si | 172.3 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 59414-23-2
Molecular Formula C8H16O2Si
Molecular Weight (g/mol) 172.3
MDL Number MFCD00008498
InChI Key SHALBPKEGDBVKK-SREVYHEPSA-N
Synonym unii-w961bik2c3, 1-methoxy-3-trimethylsiloxy-1,3-butadiene, silane, 3-methoxy-1-methylene-2-propenyl oxy trimethyl, 1-methoxy-3-trimethylsilyloxy-1,3-butadiene, silane, 3-methoxy-1-methylene-2-propen-1-yl oxy trimethyl, methyl 3-trimethylsilyloxybuta-1,3-dien-1-yl ether, 1-methoxy-3-trimethylsiloxy-1,3-butadiene, z, 1-methoxy-3-trimethylsilyloxy-1,3-butadiene unspecified, silane, 2z-3-methoxy-1-methylene-2-propen-1-yl oxy trimethyl, silane, 3-methoxy-1-methylene-2-propenyl oxy trimethyl-, z
PubChem CID 6436851
IUPAC Name [(3Z)-4-methoxybuta-1,3-dien-2-yl]oxy-trimethylsilane
SMILES COC=CC(=C)O[Si](C)(C)C

Specifications

CAS Min % 88.0
CAS Max % 100.0
Color Yellow to Brown
Density 0.8800g/mL
Boiling Point 68.0°C to 69.0°C (14.0 mmHg)
Flash Point 45°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3SiOC(=CH2)CH=CHOCH3
Refractive Index 1.4530 to 1.4550
Quantity 5 g
Fieser 06,370; 07,233; 09,303; 10,260; 11,332
Specific Gravity 0.88
Merck Index 15, 6075
Formula Weight 172.3
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material 1-Methoxy-3-(trimethylsilyloxy)-1, 3-butadiene, 90%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta
EINECSNumber 261-753-

RUO – Research Use Only