missing translation for 'onlineSavingsMsg'
Learn More

1-Methoxy-3-(trimethylsilyloxy)-1,3-butadiene 95.0+%, TCI America™

Catalog No. M07595ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M07595ML 5 mL
M075925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M07595ML Supplier TCI America Supplier No. M07595ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 59414-23-2
Molecular Formula C8H16O2Si
Molecular Weight (g/mol) 172.299
MDL Number MFCD00008498
InChI Key SHALBPKEGDBVKK-SREVYHEPSA-N
Synonym unii-w961bik2c3, 1-methoxy-3-trimethylsiloxy-1,3-butadiene, silane, 3-methoxy-1-methylene-2-propenyl oxy trimethyl, 1-methoxy-3-trimethylsilyloxy-1,3-butadiene, silane, 3-methoxy-1-methylene-2-propen-1-yl oxy trimethyl, methyl 3-trimethylsilyloxybuta-1,3-dien-1-yl ether, 1-methoxy-3-trimethylsiloxy-1,3-butadiene, z, 1-methoxy-3-trimethylsilyloxy-1,3-butadiene unspecified, silane, 2z-3-methoxy-1-methylene-2-propen-1-yl oxy trimethyl, silane, 3-methoxy-1-methylene-2-propenyl oxy trimethyl-, z
PubChem CID 6436851
IUPAC Name [(3Z)-4-methoxybuta-1,3-dien-2-yl]oxy-trimethylsilane
SMILES COC=CC(=C)O[Si](C)(C)C

Specifications

Color Colorless
Quantity 5 mL
UN Number 1993
Formula Weight 172.30
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Methoxy-3-(trimethylsilyloxy)-1,3-butadiene
TSCA Yes
Recommended Storage Refrigerator