accessibility menu, dialog, popup

UserName

CAS RN 591-78-6

IUPAC Name: hexan-2-one
Synonyms: hexanone MNBK butyl methyl ketone (substance) hexan 2 one MBK N-butyl methyl ketone 2 hexanone methyl n butyl ketone
Molecular Weight (g/mol): 100.16
Molecular Formula: C6H12O
InChi Key: QQZOPKMRPOGIEB-UHFFFAOYSA-N
SMILES: CCCCC(C)=O
Percent Purity 
  • (3)
  • (1)
  • (7)
Boiling Point 
  • (4)
  • (6)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (1)
  • (3)
  • (7)
  • (3)
  • (1)
  • (7)
  • (4)
  • (6)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (6)
  • (1)