accessibility menu, dialog, popup

UserName

ClH3CNH3CCH3OHO

CAS RN 590-46-5

IUPAC Name: (carboxymethyl)trimethylazanium chloride
Synonyms: acidine
Molecular Weight (g/mol): 153.61
Molecular Formula: C5H12ClNO2
InChi Key: HOPSCVCBEOCPJZ-UHFFFAOYSA-N
SMILES: [Cl-].C[N+](C)(C)CC(O)=O
Percent Purity 
  • (2)
  • (4)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Grade 
  • (2)
  • (1)
  • (4)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (4)
  • (1)
  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (2)