missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Betaine hydrochloride, MilliporeSigma™ Supelco™

Catalog No. 111001972
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001972 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001972 Supplier Merck Emd Millipore Supplier No. 9091050MG

missing translation for 'validMainframeMsg'

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.

Specifications

CAS 590-46-5
Product Line TraceCERT
Linear Formula (CH3)3 N+CH2 COOH · Cl-
MDL Number MFCD00011903
Melting Point 241°C to 242°C
Synonym (Carboxymethyl)trimethylammonium hydrochloride
Type Certified Reference Material
Quantity 50 mg
Formula Weight 153.61
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC