accessibility menu, dialog, popup

UserName

CAS RN 590-02-3

H3COOCl

CAS RN 590-02-3

IUPAC Name: butyl 2-chloroacetate
Synonyms: butyl 2-chloroacetate
Molecular Weight (g/mol): 150.6
Molecular Formula: C6H11ClO2
InChi Key: YJRGMUWRPCPLNH-UHFFFAOYSA-N
SMILES: CCCCOC(=O)CCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results