missing translation for 'onlineSavingsMsg'
Learn More

Butyl Chloroacetate 98.0+%, TCI America™

Catalog No. C009125ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C009125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C009125ML Supplier TCI America Supplier No. C009125ML
Only null left

Chemical Identifiers

CAS 590-02-3
Molecular Formula C6H11ClO2
Molecular Weight (g/mol) 150.60
MDL Number MFCD00018925
InChI Key YJRGMUWRPCPLNH-UHFFFAOYSA-N
Synonym Chloroacetic Acid Butyl Ester
PubChem CID 11530
IUPAC Name butyl 2-chloroacetate
SMILES CCCCOC(=O)CCl

Specifications

Color Colorless
Boiling Point 183°C
Quantity 25 mL
Formula Weight 150.60
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyl Chloroacetate
EINECSNumber (2)-1149
TSCA Yes