accessibility menu, dialog, popup

UserName

H3CHNNH2HCl

CAS RN 58711-02-7

IUPAC Name: (2-ethylphenyl)hydrazine hydrochloride
Synonyms: (2-ethylphenyl)hydrazine hydrochloride
Molecular Weight (g/mol): 172.66
Molecular Formula: C8H13ClN2
InChi Key: HBHPTOKYVGZBAJ-UHFFFAOYSA-N
SMILES: Cl.CCC1=CC=CC=C1NN
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results