accessibility menu, dialog, popup

UserName

F

CAS RN 587-79-1

IUPAC Name: 1-benzyl-4-fluorobenzene
Synonyms: 1-benzyl-4-fluorobenzene
Molecular Weight (g/mol): 186.23
Molecular Formula: C13H11F
InChi Key: ADCBAIUWZPOIMC-UHFFFAOYSA-N
SMILES: FC1=CC=C(CC2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results