accessibility menu, dialog, popup

UserName

CAS RN 586-77-6

IUPAC Name: 4-bromo-N,N-dimethylaniline
Synonyms: 4-bromo-N,N-dimethylaniline
Molecular Weight (g/mol): 200.08
Molecular Formula: C8H10BrN
InChi Key: XYZWMVYYUIMRIZ-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(Br)C=C1
Boiling Point 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (3)
  • (3)
  • (1)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)