missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-N,N-dimethylaniline, 99%

Catalog No. AC222211000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC222211000 100 g Plastic bottle
AC222210250 25 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC222211000 Supplier Thermo Scientific Chemicals Supplier No. 222211000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 586-77-6 | C8H10BrN | 200.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 586-77-6
Molecular Formula C8H10BrN
Molecular Weight (g/mol) 200.08
MDL Number MFCD00000093
InChI Key XYZWMVYYUIMRIZ-UHFFFAOYSA-N
Synonym n,n-dimethyl-p-bromoaniline, 4-dimethylaminobromobenzene, p-bromo-n,n-dimethylaniline, benzenamine, 4-bromo-n,n-dimethyl, n,n-dimethyl-4-bromoaniline, p-dimethylamino phenyl bromide, p-dimethylaminobromobenzene, 1-bromo-4-dimethylamino benzene, p-bromo dimethylamino benzene, p-n,n-dimethylaminobromobenzene
PubChem CID 11465
IUPAC Name 4-bromo-N,N-dimethylaniline
SMILES CN(C)C1=CC=C(Br)C=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 53°C to 56°C
Color Blue or Gray to White
Boiling Point 264°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula BrC6H4N(CH3)2
Quantity 100 g
Beilstein 12, 637
Solubility Information Solubility in water: insoluble. Other solubilities: methanol 100g/L
Formula Weight 200.08
Percent Purity 99%
Physical Form Crystalline Solid
Chemical Name or Material 4-Bromo-N, N-dimethylaniline, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Use personal protective equipment as required.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye
EINECSNumber 209-582-2
RTECSNumber BW9300000
TSCA TSCA

RUO – Research Use Only