missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-N,N-dimethylaniline 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 54°C |
| Color | White |
| Boiling Point | 264°C |
| Quantity | 500 g |
| Formula Weight | 200.08 |
| Percent Purity | ≥98.0% (GC) |
| Grade | Not Available |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Bromo-N,N-dimethylaniline |
Safety and Handling
| RTECSNumber | BW9300000 |
| TSCA | Yes |