accessibility menu, dialog, popup

UserName

HOOOH

CAS RN 58574-03-1

IUPAC Name: 4'-hydroxy-[1,1'-biphenyl]-4-carboxylic acid
Synonyms: 4'-hydroxy-[1,1'-biphenyl]-4-carboxylic acid
Molecular Weight (g/mol): 214.22
Molecular Formula: C13H10O3
InChi Key: JTGCXYYDAVPSFD-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1)C1=CC=C(O)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)