accessibility menu, dialog, popup

UserName

CAS RN 585-70-6

IUPAC Name: 5-bromofuran-2-carboxylic acid
Synonyms: 5-bromofuran-2-carboxylic acid
Molecular Weight (g/mol): 190.98
Molecular Formula: C5H3BrO3
InChi Key: YVTQHZDUDUCGRD-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(Br)O1
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (1)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)