accessibility menu, dialog, popup

UserName

ONOBrNOO

CAS RN 584-48-5

IUPAC Name: 1-bromo-2,4-dinitrobenzene
Synonyms: DNBB
Molecular Weight (g/mol): 247
Molecular Formula: C6H3BrN2O4
InChi Key: PBOPJYORIDJAFE-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(Br)C(=C1)[N+]([O-])=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)