accessibility menu, dialog, popup

UserName

H3CSOOONa

CAS RN 5802-89-1

IUPAC Name: sodium tridecane-1-sulfonate
Synonyms: sodium tridecane-1-sulfonate
Molecular Weight (g/mol): 286.41
Molecular Formula: C13H27NaO3S
InChi Key: CACJZDMMUHMEBN-UHFFFAOYSA-M
SMILES: [Na+].CCCCCCCCCCCCCS([O-])(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results