accessibility menu, dialog, popup

UserName

CAS RN 579-74-8

IUPAC Name: 1-(2-methoxyphenyl)ethan-1-one
Synonyms: ethanone, 1-(2-methoxyphenyl)-
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: DWPLEOPKBWNPQV-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1C(C)=O
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)