accessibility menu, dialog, popup

UserName

CAS RN 579-11-3

CH3ONCl

CAS RN 579-11-3

IUPAC Name: N-chloro-N-phenylacetamide
Synonyms: N-chloro-N-phenylacetamide
Molecular Weight (g/mol): 169.61
Molecular Formula: C8H8ClNO
InChi Key: QNVKMXGRFVLMBM-UHFFFAOYSA-N
SMILES: CC(=O)N(Cl)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results