missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Chloroacetanilide 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 92°C |
| Color | White-Yellow |
| Quantity | 1 g |
| UN Number | 3261 |
| Formula Weight | 169.61 |
| Percent Purity | ≥98.0% (N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | N-Chloroacetanilide |
Safety and Handling
| RTECSNumber | AE0350000 |
| TSCA | No |
| Recommended Storage | Freezer |