accessibility menu, dialog, popup

UserName

CH3NBrOHOO(E)

CAS RN 5785-70-6

IUPAC Name: (4E)-5-(4-bromophenyl)-4-[hydroxy(phenyl)methylidene]-1-methylpyrrolidine-2,3-dione
Synonyms: (4E)-5-(4-bromophenyl)-4-[hydroxy(phenyl)methylidene]-1-methylpyrrolidine-2,3-dione
Molecular Weight (g/mol): 372.22
Molecular Formula: C18H14BrNO3
InChi Key: ORVACIWFPOGOSU-JQIJEIRANA-N
SMILES: CN1C(\C(=C(/O)C2=CC=CC=C2)C(=O)C1=O)C1=CC=C(Br)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results