accessibility menu, dialog, popup

UserName

H2OH2OH2OH2OCaCaCaOHOOOOOOOHOOOOOO

CAS RN 5785-44-4

IUPAC Name: tricalcium bis(2-hydroxypropane-1,2,3-tricarboxylate) tetrahydrate
Synonyms: tricalcium dicitrate tetrahydrate
Molecular Weight (g/mol): 570.49
Molecular Formula: C12H18Ca3O18
InChi Key: LNIZKKFWMDARJV-UHFFFAOYSA-H
SMILES: O.O.O.O.[Ca++].[Ca++].[Ca++].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O.OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Molecular Weight (g/mol) 
  • (7)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (4)
  • (3)
  • (2)
Grade 
  • (2)
  • (4)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (7)
  • (3)
  • (4)
  • (4)
  • (7)
  • (2)
  • (2)
  • (2)
  • (4)
  • (3)
  • (2)
  • (2)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)