accessibility menu, dialog, popup

UserName

CAS RN 578-67-6

IUPAC Name: quinolin-1-ium-5-olate
Synonyms: quinolin-1-ium-5-olate
Molecular Weight (g/mol): 145.16
Molecular Formula: C9H7NO
InChi Key: GYESAYHWISMZOK-UHFFFAOYSA-N
SMILES: [O-]C1=C2C=CC=[NH+]C2=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)