missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxyquinoline, 97%

Catalog No. AC156430050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC156430050 5 g Glass bottle
AC156430010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC156430050 Supplier Thermo Scientific Chemicals Supplier No. 156430050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 578-67-6 | C9H7NO | 145.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 578-67-6
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD00006792
InChI Key GVNQVWJYDXOLST-UHFFFAOYSA-N
Synonym 5-hydroxyquinoline, quinolin-5-ol, 5-quinolinol, 5-chinolinol, ccris 4330, gyesayhwismzok-uhfffaoysa-n, 5-hydroxyquinoline 5-quinolinol, quinolin-5-one, 5-hyroxyquinoline, 5-hydroxy quinoline
PubChem CID 11360
IUPAC Name 1H-quinolin-5-one
SMILES C1=CC(=O)C2=CC=CNC2=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 223°C to 226°C
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 145.16
Percent Purity 97%
Physical Form Solution
Chemical Name or Material 5-Hydroxyquinoline
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 209-428-4

RUO – Research Use Only