accessibility menu, dialog, popup

UserName

H3CH2N

CAS RN 578-54-1

IUPAC Name: 2-ethylaniline
Synonyms: O-ethylaniline
Molecular Weight (g/mol): 121.18
Molecular Formula: C8H11N
InChi Key: MLPVBIWIRCKMJV-UHFFFAOYSA-N
SMILES: CCC1=CC=CC=C1N
Boiling Point 
  • (1)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (1)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)