missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylaniline, 97%

Catalog No. AAA1498422
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1498422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1498422 Supplier Thermo Scientific Chemicals Supplier No. A1498422
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 578-54-1 | C8H11N | 121.183 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 578-54-1
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.183
MDL Number MFCD00007751
InChI Key MLPVBIWIRCKMJV-UHFFFAOYSA-N
Synonym o-ethylaniline, benzenamine, 2-ethyl, aniline, o-ethyl, 2-ethylbenzenamine, o-aminoethylbenzene, aniline, 2-ethyl, 2-ethyl aniline, 2-ethylphenylamine, unii-gr557n47k6, ccris 2858
PubChem CID 11357
IUPAC Name 2-ethylaniline
SMILES CCC1=CC=CC=C1N

Specifications

Melting Point -47°C
Density 0.983
Boiling Point 208°C to 210°C
Flash Point 91°C (196°F)
Odor Characteristic
Refractive Index 1.56
Quantity 100 g
UN Number UN2273
Beilstein 1841268
Formula Weight 121.18
Percent Purity 97%
Chemical Name or Material 2-Ethylaniline
Show More Show Less
Hazard Category H227-H301+H311+H331-H373
Hazard Statement GHS H Statement
H331-H373-H227-H302-H312
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P210-P233-P240-P243-P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P362-P374-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: 2-ETHYLANILINE
EINECSNumber 209-424-2
RTECSNumber BX9800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only