missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylaniline 98.0+%, TCI America™

Catalog No. E0060500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0060500ML 500 mL
E006025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0060500ML Supplier TCI America Supplier No. E0060500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 578-54-1
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.183
MDL Number MFCD00007751
InChI Key MLPVBIWIRCKMJV-UHFFFAOYSA-N
Synonym o-ethylaniline, benzenamine, 2-ethyl, aniline, o-ethyl, 2-ethylbenzenamine, o-aminoethylbenzene, aniline, 2-ethyl, 2-ethyl aniline, 2-ethylphenylamine, unii-gr557n47k6, ccris 2858
PubChem CID 11357
IUPAC Name 2-ethylaniline
SMILES CCC1=CC=CC=C1N

Specifications

Melting Point -44°C
Color Yellow
Boiling Point 216°C
Quantity 500 mL
UN Number 2273
Formula Weight 121.18
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Ethylaniline
EINECSNumber (3)-0206
RTECSNumber BX9800000
TSCA Yes