accessibility menu, dialog, popup

UserName

CH3OHBrBrBrBr

CAS RN 576-55-6

IUPAC Name: 2,3,4,5-tetrabromo-6-methylphenol
Synonyms: 2,3,4,5-tetrabromo-6-methylphenol
Molecular Weight (g/mol): 423.72
Molecular Formula: C7H4Br4O
InChi Key: GGIDUULRWQOXLR-UHFFFAOYSA-N
SMILES: CC1=C(O)C(Br)=C(Br)C(Br)=C1Br
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results