accessibility menu, dialog, popup

UserName

HClCH3CH3H3CNHHO

CAS RN 57497-39-9

IUPAC Name: hydrogen N-tert-butylhydroxylamine chloride
Synonyms: hydrogen N-tert-butylhydroxylamine chloride
Molecular Weight (g/mol): 125.6
Molecular Formula: C4H12ClNO
InChi Key: DCSATTBHEMKGIP-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC(C)(C)NO
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)