missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butyl)hydroxylamine Hydrochloride 98.0+%, TCI America™

Catalog No. B17005G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17005G 5 g
B17001G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B17005G Supplier TCI America Supplier No. B17005G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 57497-39-9
Molecular Formula C4H12ClNO
Molecular Weight (g/mol) 125.596
MDL Number MFCD00012598
InChI Key DCSATTBHEMKGIP-UHFFFAOYSA-N
Synonym n-tert-butyl hydroxylamine hydrochloride, n-tert-butyl hydroxylamine hcl, n-tert-butylhydroxylamine hydrochloride, n-1,1-dimethylethyl hydroxylamine hydrochloride, 2-propanamine, n-hydroxy-2-methyl-, hydrochloride, 2-methylprop-2-ylhydroxylamine, chloride, soyheadhjuup@, acmc-1b1z0, t-butylhydroxylamine hydrochloride
PubChem CID 2777896
IUPAC Name N-tert-butylhydroxylamine;hydrochloride
SMILES CC(C)(C)NO.Cl

Specifications

Melting Point 185°C
Color White
Quantity 5 g
Formula Weight 125.60
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butyl)hydroxylamine Hydrochloride