CAS RN 5731-10-2
IUPAC Name:
4'-fluoro-[1,1'-biphenyl]-4-carboxylate
Synonyms:
4'-fluoro-[1,1'-biphenyl]-4-carboxylate
Molecular Weight (g/mol):
215.2
Molecular Formula:
C13H8FO2
InChi Key:
LXWNTLBMNCXRQN-UHFFFAOYSA-M
SMILES:
[O-]C(=O)C1=CC=C(C=C1)C1=CC=C(F)C=C1
1
–
1
of
1
results
1
Promotions Available
4'-Fluorobiphenyl-4-carboxylic acid, 96%
4'-Fluorobiphenyl-4-carboxylic acid, 96%
CAS: 5731-10-2 Molecular Formula: C13H8FO2 Molecular Weight (g/mol): 215.20 MDL Number: MFCD01631909 InChI Key: LXWNTLBMNCXRQN-UHFFFAOYSA-M Synonym: 4-4-fluorophenyl benzoic acid,4'-fluoro-biphenyl-4-carboxylic acid,4'-fluorobiphenyl-4-carboxylic acid,4'-fluoro-1,1'-biphenyl-4-carboxylic acid,4-biphenyl-4'-fluoro-carboxylic acid,4'-fluoro-biphenyl-4-carboxylic acid,4-fluoro-biphenyl-4-carboxylic acid,4'-fluoro 1,1'-biphenyl-4-carboxylic acid,4fc PubChem CID: 2782689 IUPAC Name: 4-(4-fluorophenyl)benzoic acid SMILES: C1=CC(=CC=C1C2=CC=C(C=C2)F)C(=O)O