accessibility menu, dialog, popup

UserName

CAS RN 572-83-8

IUPAC Name: 2-bromo-9,10-dihydroanthracene-9,10-dione
Synonyms: 2-bromoanthracene-9,10-dione
Molecular Weight (g/mol): 287.11
Molecular Formula: C14H7BrO2
InChi Key: VTSDGYDTWADUJQ-UHFFFAOYSA-N
SMILES: BrC1=CC=C2C(=O)C3=CC=CC=C3C(=O)C2=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)