missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoanthraquinone 96.0+%, TCI America™

Catalog No. B26895G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B26895G 5 g
B26891G 1 g
B268925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B26895G Supplier TCI America Supplier No. B26895G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 572-83-8
Molecular Formula C14H7BrO2
Molecular Weight (g/mol) 287.112
InChI Key VTSDGYDTWADUJQ-UHFFFAOYSA-N
PubChem CID 221013
IUPAC Name 2-bromoanthracene-9,10-dione
SMILES C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C=C(C=C3)Br

Specifications

Melting Point 208°C
Quantity 5 g
Formula Weight 287.11
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromoanthraquinone
RTECSNumber CB5950000
TSCA No