accessibility menu, dialog, popup

UserName

HO(R)O

CAS RN 57044-25-4

IUPAC Name: [(2R)-oxiran-2-yl]methanol
Synonyms: (+)-glycidol
Molecular Weight (g/mol): 74.08
Molecular Formula: C3H6O2
InChi Key: CTKINSOISVBQLD-GSVOUGTGSA-N
SMILES: OC[C@@H]1CO1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results