missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-Glycidol 98.0+%, TCI America™

Catalog No. G03635G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G03635G 5 g
G036325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G03635G Supplier TCI America Supplier No. G03635G

Chemical Identifiers

CAS 57044-25-4
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.079
MDL Number MFCD00074873
InChI Key CTKINSOISVBQLD-GSVOUGTGSA-N
Synonym (R)-(+)-1-Hydroxy-2,3-epoxypropane, (R)-(+)-2-(Hydroxymethyl)oxirane
PubChem CID 5460455
ChEBI CHEBI:18664
IUPAC Name [(2R)-oxiran-2-yl]methanol
SMILES C1C(O1)CO

Specifications

Color Yellow
Boiling Point 57°C
Quantity 5 g
UN Number 2810
Formula Weight 74.08
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-(+)-Glycidol
EINECSNumber (2)-2389
RTECSNumber RR0508000
TSCA No
Recommended Storage Freezer