accessibility menu, dialog, popup

UserName

H3COOOONNH2NH3CH3SOOO(E/Z)

CAS RN 56974-61-9

IUPAC Name: ethyl 4-[(6-{[amino(azaniumyl)methylidene]amino}hexanoyl)oxy]benzoate methanesulfonate
Synonyms: ethyl 4-[(6-{[amino(ammonio)methylidene]amino}hexanoyl)oxy]benzoate mesylate
Molecular Weight (g/mol): 417.48
Molecular Formula: C17H27N3O7S
InChi Key: DNTNDFLIKUKKOC-UHFFFAOYSA-N
SMILES: CS([O-])(=O)=O.CCOC(=O)C1=CC=C(OC(=O)CCCCCN=C(N)[NH3+])C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results