accessibility menu, dialog, popup

UserName

H3CCH3OClOCl

CAS RN 5659-93-8

IUPAC Name: dimethylpropanedioyl dichloride
Synonyms: dimethylpropanedioyl dichloride
Molecular Weight (g/mol): 169
Molecular Formula: C5H6Cl2O2
InChi Key: CJXQAYQWVNXIQE-UHFFFAOYSA-N
SMILES: CC(C)(C(Cl)=O)C(Cl)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results