missing translation for 'onlineSavingsMsg'
Learn More

Dimethylmalonyl Dichloride 98.0+%, TCI America™

Catalog No. D27235G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27235G 5 g
D272325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D27235G Supplier TCI America Supplier No. D27235G
Only null left

Chemical Identifiers

CAS 5659-93-8
Molecular Formula C5H6Cl2O2
Molecular Weight (g/mol) 169.001
MDL Number MFCD00192081
InChI Key CJXQAYQWVNXIQE-UHFFFAOYSA-N
PubChem CID 4523180
IUPAC Name 2,2-dimethylpropanedioyl dichloride
SMILES CC(C)(C(=O)Cl)C(=O)Cl

Specifications

Color Yellow
Quantity 5 g
UN Number 2920
Formula Weight 169.00
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Dimethylmalonyl Dichloride