accessibility menu, dialog, popup

UserName

CH3H3CBrOHO

CAS RN 565-74-2

IUPAC Name: 2-bromo-3-methylbutanoic acid
Synonyms: 2-bromo-3-methyl-butanoic acid
Molecular Weight (g/mol): 181.03
Molecular Formula: C5H9BrO2
InChi Key: UEBARDWJXBGYEJ-UHFFFAOYNA-N
SMILES: CC(C)C(Br)C(O)=O
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)