missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-methylbutyric Acid 90.0+%, TCI America™

Catalog No. B066825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B066825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B066825G Supplier TCI America Supplier No. B066825G

Chemical Identifiers

CAS 565-74-2
Molecular Formula C5H9BrO2
Molecular Weight (g/mol) 181.029
MDL Number MFCD00004210
InChI Key UEBARDWJXBGYEJ-UHFFFAOYSA-N
Synonym 2-bromo-3-methylbutyric acid, 2-bromoisovaleric acid, butanoic acid, 2-bromo-3-methyl, alpha-bromoisovaleric acid, butyric acid, 2-bromo-3-methyl, 2-bromo-3-methyl-butanoic acid, .alpha.-bromoisovaleric acid, +/--2-bromo-3-methylbutyric acid, bromisovaleriansaure, acmc-209wpb
PubChem CID 11268
IUPAC Name 2-bromo-3-methylbutanoic acid
SMILES CC(C)C(C(=O)O)Br

Specifications

Color White-Yellow
Boiling Point 230°C
Quantity 25 g
UN Number 3261
Formula Weight 181.03
Percent Purity ≥90.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Bromo-3-methylbutyric Acid
RTECSNumber ES8055000
TSCA Yes