accessibility menu, dialog, popup

UserName

CAS RN 565-64-0

IUPAC Name: 2,3-dichloropropanoic acid
Synonyms: 2,3-dichloropropionic acid
Molecular Weight (g/mol): 142.96
Molecular Formula: C3H4Cl2O2
InChi Key: GKFWNPPZHDYVLI-UHFFFAOYNA-N
SMILES: OC(=O)C(Cl)CCl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results