missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloropropionic Acid 98.0+%, TCI America™

Catalog No. D04075G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D04075G 5 g
D040725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D04075G Supplier TCI America Supplier No. D04075G

Chemical Identifiers

CAS 565-64-0
Molecular Formula C3H4Cl2O2
Molecular Weight (g/mol) 142.963
MDL Number MFCD00021695
InChI Key GKFWNPPZHDYVLI-UHFFFAOYSA-N
PubChem CID 11263
IUPAC Name 2,3-dichloropropanoic acid
SMILES C(C(C(=O)O)Cl)Cl

Specifications

Melting Point 52°C
Color White-Yellow
Boiling Point 210°C
Quantity 5 g
UN Number 2923
Formula Weight 142.96
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dichloropropionic Acid
RTECSNumber UF0700000
TSCA No